Learn More
Medchemexpress LLC Kw-8232 | 217813-15-5 | 98.1% | 705.26 g·mol⁻¹ | C37H41ClN4O6S | 10 MG

Supplier: Medchemexpress LLC HY100304A10MG
KW-8232 is a small-molecule research compound reported as an orally active anti-osteoporotic agent that reduces prostaglandin E2 (PGE2) biosynthesis. It has been evaluated in preclinical bone-loss studies and has reported in vitro antiviral activity against SARS-CoV-2. Supplied for research use with supporting analytical documentation.
- Orally active anti-osteoporotic agent that reduces PGE2 biosynthesis.
- Reported in vitro antiviral activity against SARS-CoV-2 (EC50 ≈ 1.2 μM).
- High purity, 98.1%.
- Molecular weight 705.26 g·mol⁻¹; formula C37H41ClN4O6S.
- Soluble in DMSO at 125 mg/mL; ultrasonic assistance may be required.
- Available in multiple laboratory pack sizes and solution formats.
- Supplier documentation available: data sheet, certificate of analysis, and safety data sheet.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.