Learn More
Medchemexpress LLC Kw-8232 | 217813-15-5 | 98.1% | 705.26 g/mol | C37H41ClN4O6S | 50 MG
SDP
Catalog No. 5000214126
Encompass_Preferred
KW-8232 is an orally active anti-osteoporotic small molecule that reduces prostaglandin E2 (PGE2) biosynthesis in osteoblastic cells and has reported antiviral activity against SARS-CoV-2 (EC50 ≈1.2 μM). Supplied as a solid for research use with reported high purity and recommended storage conditions.
Catalog No. 50-002-14126 Supplier Medchemexpress LLC Supplier No. HY100304A50MG
KW-8232 is an orally active anti-osteoporotic small molecule that reduces prostaglandin E2 (PGE2) biosynthesis in osteoblastic cells and has reported antiviral activity against SARS-CoV-2 (EC50 ≈1.2 μM). Supplied as a solid for research use with reported high purity and recommended storage conditions.