Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Kw-8232 methanesulfonate | 217813-15-5 | 98.1% | 705.26 g·mol⁻¹ | C37H41ClN4O6S | 100 MG
SDP

Catalog No. 5000214009
Encompass_Preferred

KW-8232 methanesulfonate is an orally active anti-osteoporotic small molecule that reduces prostaglandin E2 (PGE2) biosynthesis in osteoblastic cells and has been shown to inhibit bone loss in preclinical studies. It is supplied for research use in solid form and as a 10 mM solution in DMSO.

  • Orally active anti-osteoporotic agent.
  • Reduces prostaglandin E2 (PGE2) biosynthesis in osteoblastic cells.
  • Demonstrated inhibition of bone loss in preclinical models.
  • High purity (98.1%).
  • Molecular weight 705.26 g·mol⁻¹; formula C37H41ClN4O6S.
  • Available as solid and as a 10 mM solution in DMSO for in vitro and in vivo studies.

Catalog No. 50-002-14009 Supplier Medchemexpress LLC Supplier No. HY100304A100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

KW-8232 methanesulfonate is an orally active anti-osteoporotic small molecule that reduces prostaglandin E2 (PGE2) biosynthesis in osteoblastic cells and has been shown to inhibit bone loss in preclinical studies. It is supplied for research use in solid form and as a 10 mM solution in DMSO.

  • Orally active anti-osteoporotic agent.
  • Reduces prostaglandin E2 (PGE2) biosynthesis in osteoblastic cells.
  • Demonstrated inhibition of bone loss in preclinical models.
  • High purity (98.1%).
  • Molecular weight 705.26 g·mol⁻¹; formula C37H41ClN4O6S.
  • Available as solid and as a 10 mM solution in DMSO for in vitro and in vivo studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.