Learn More
Abcam KW2449, 5MG
Catalog No. 502682925
Encompass_Preferred
MW 332.4 Da, Purity >98%. Multi targeted inhibitor of FLT3 ( with an IC₅₀ of 6.6nM), ABL, ABL-T315I, and Aurora kinase. Tumor growth inhibition in FLT3-mutated xenograft model in vitro. Shows a potent and significant antitumor effect in a dose-dependent manner in MOLM-13 tumour xenograft model in vivo..
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-268-2925 Supplier Abcam Supplier No. AB2735965MG
MW 332.4 Da, Purity >98%. Multi targeted inhibitor of FLT3 ( with an IC₅₀ of 6.6nM), ABL, ABL-T315I, and Aurora kinase. Tumor growth inhibition in FLT3-mutated xenograft model in vitro. Shows a potent and significant antitumor effect in a dose-dependent manner in MOLM-13 tumour xenograft model in vivo..
The product is subject to the following: Abcam Restricted Use Statement