Learn More
Showa Denko America Inc KW404-4F
Supplier:  Showa Denko America Inc F6989203
Encompass_Preferred
High performance SEC GFC silica packed KW-400 series, reduced packing material particle size enhances column performance, three to four-fold higher sensitivity than KW-800 series, KW405-4F is applicable analyzing samples with molecular weight above 1,000,000, fulfills USP L20, L33, and L59 requirements, MW range (pullulan) = 20,000 to 300,000 (estimated) Daltons, dimensions: 4.6 mm ID x 300 mm L
Catalog No. 50-161-3284