Learn More
World Precision Instrument Kwik-Gard
SDP
Catalog No. 502538733
Encompass
Kwik-Gard is specially packaged Sylgard184 silicone for quicker and easier application, eliminating the messy procedure of preparing the mixture before application. Its special cartridge controls the precise mixing ratio to ensure proper curing. The disposable tip mixes resin and hardener as they are dispensed. Since no air is introduced during mixing, the resin does not need degassing for most applications. The mixed silicone is applied directly to the site, reducing preparation time and material waste. Each Kwik-Gard cartridge contains 37 mL of resin and hardener. The dispensing tip has a dead volume of 0.75 mL.
Catalog No. 50-253-8733 Supplier World Precision Instrument Supplier No. KWIKGARD
Kwik-Gard is specially packaged Sylgard184 silicone for quicker and easier application, eliminating the messy procedure of preparing the mixture before application. Its special cartridge controls the precise mixing ratio to ensure proper curing. The disposable tip mixes resin and hardener as they are dispensed. Since no air is introduced during mixing, the resin does not need degassing for most applications. The mixed silicone is applied directly to the site, reducing preparation time and material waste. Each Kwik-Gard cartridge contains 37 mL of resin and hardener. The dispensing tip has a dead volume of 0.75 mL.