Learn More
Micro Biologics Inc 3P Klebsiella pneumoniae derived from NCTC 13439, 2 pack
SDP
Catalog No. NC1766112
Encompass_Preferred
Strain Characteristics: blaVIM-1gene, Metallo-beta-lactamase positive | Antibiotic/Drug Resistant Strains: Metallo-beta-lactamase | Test Method: Antimicrobial Susceptibility Testing, Microbial Identification
This item is not returnable. View return policy
Catalog No. NC1766112 Supplier Micro Biologics Inc 3P Supplier No. 01245P
Strain Characteristics: blaVIM-1gene, Metallo-beta-lactamase positive | Antibiotic/Drug Resistant Strains: Metallo-beta-lactamase | Test Method: Antimicrobial Susceptibility Testing, Microbial Identification