Learn More
MSC Zep Solvent Degreaser, Fast-Evaporating, Residue-Free, Aerosol, 22 oz
Catalog No. 502870444
Encompass
Zep Kwik-Solv is a clear, colorless solvent degreaser designed for fast evaporation and residue-free cleaning. It effectively removes tough grease, oils, grime, and other oil-based contaminants from various surfaces. This product ensures heavy-duty cleaning without leaving residue, allowing equipment to be returned to use promptly.
  • Quickly penetrates and removes oils, greases, and soils from surfaces
  • Contains no detergents, builders, or chlorinated solvents
  • Evaporates completely without leaving a residue
  • Suited for use on circuit boards, electrical contacts, electric motors, machinery parts, and electronic equipment
Catalog No. 50-287-0444 Supplier MSC Supplier No. 11395183
Zep Kwik-Solv is a clear, colorless solvent degreaser designed for fast evaporation and residue-free cleaning. It effectively removes tough grease, oils, grime, and other oil-based contaminants from various surfaces. This product ensures heavy-duty cleaning without leaving residue, allowing equipment to be returned to use promptly.
  • Quickly penetrates and removes oils, greases, and soils from surfaces
  • Contains no detergents, builders, or chlorinated solvents
  • Evaporates completely without leaving a residue
  • Suited for use on circuit boards, electrical contacts, electric motors, machinery parts, and electronic equipment