Learn More
Health Care Logistics Inc KwikDose™ Oral Dispensers with Tip Caps, 0.5mL, Amber, 100/pack
SDP
Catalog No. NC3957380
Encompass
Familiar, Trusted Design - Oral tip is designed to discourage wrong-route medication delivery.Safer Dispensing - High-quality plunger easily draws and dispenses oral medication.Colors - Clear and AmberSizes - 0.5mL, 1mL, 3mL, 5mL, 10mL and 20mLQuantity: 100 per package; 500 per case. Cases include 5 inner packages of 100. All packages include matching number of tip caps, bagged separately.Note - Oral Dispensers are intended for immediate use only.
Catalog No. NC3957380 Supplier Health Care Logistics Inc Supplier No. 21184A
Familiar, Trusted Design - Oral tip is designed to discourage wrong-route medication delivery.Safer Dispensing - High-quality plunger easily draws and dispenses oral medication.Colors - Clear and AmberSizes - 0.5mL, 1mL, 3mL, 5mL, 10mL and 20mLQuantity: 100 per package; 500 per case. Cases include 5 inner packages of 100. All packages include matching number of tip caps, bagged separately.Note - Oral Dispensers are intended for immediate use only.