Learn More
Selleck Chemical LLC KX1-004
SDP
Catalog No. 5000002985
Encompass_Preferred
KX1-004 is a non-ATP competitive inhibitor of Src protein tyrosine kinase (Src-PTK IC50 40 M)
Catalog No. 50-000-02985 Supplier Selleck Chemical LLC Supplier No. S65002MG
KX1-004 is a non-ATP competitive inhibitor of Src protein tyrosine kinase (Src-PTK IC50 40 M)