Learn More
Medchemexpress LLC KY-02327 acetate | 2989396-34-9 | 99.2% | 433.50 g/mol | C22H31N3O6 | 10MM 1ML
SDP
Catalog No. 5000236972
Encompass_Preferred
KY-02327 acetate is a small-molecule inhibitor of the Dishevelled (Dvl)-CXXC5 interaction that activates the Wnt/β-catenin signaling pathway. It is supplied as a solution in DMSO for cell biology and biochemical assays and is commonly used to study osteoblast differentiation and pathway modulation. The compound has formula C22H31N3O6, molecular weight 433.50 g/mol, and reported purity ~99.2%.
  • Provided as a ready-to-use solution for rapid experimental setup.
  • Reported high purity to minimize impurity-related effects.
  • Suitable for Wnt pathway and osteoblast differentiation studies.
  • Available in solid form for custom formulation or storage.
  • Compact packaging reduces storage footprint and waste.
  • Stable under recommended storage conditions for reliable results.
Catalog No. 50-002-36972 Supplier Medchemexpress LLC Supplier No. HY124156A10MM 1ML
KY-02327 acetate is a small-molecule inhibitor of the Dishevelled (Dvl)-CXXC5 interaction that activates the Wnt/β-catenin signaling pathway. It is supplied as a solution in DMSO for cell biology and biochemical assays and is commonly used to study osteoblast differentiation and pathway modulation. The compound has formula C22H31N3O6, molecular weight 433.50 g/mol, and reported purity ~99.2%.
  • Provided as a ready-to-use solution for rapid experimental setup.
  • Reported high purity to minimize impurity-related effects.
  • Suitable for Wnt pathway and osteoblast differentiation studies.
  • Available in solid form for custom formulation or storage.
  • Compact packaging reduces storage footprint and waste.
  • Stable under recommended storage conditions for reliable results.