Learn More
Medchemexpress LLC HY-124745 5mg Medchemexpress, KY-05009 CAS:1228280-29-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1247455MG
Encompass_Preferred
Medchemexpress, HY-124745 5mg KY-05009 CAS:1228280-29-2 KY-05009 is an ATP-competitive Traf2- and Nck-interacting kinase (TNIK) inhibitor with a Ki of 100 nM. KY-05009 pharmacologically inhibits TGF-β1-induced epithelial-to-mesenchymal transition (EMT) in human lung adenocarcinoma cells. KY-05009 inhibits the protein expression of TNIK and transcriptional activity of Wnt target genes and induces apoptosis in cancer cells. KY-05009 exerts anti-cancer activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0004