Learn More
Medchemexpress LLC HY-120327 10mg Medchemexpress, KY-226 CAS:1621673-53-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY12032710MG
Encompass_Preferred
Medchemexpress, HY-120327 10mg KY-226 CAS:1621673-53-7 KY-226 is a potent, selective, orally active and allosteric protein tyrosine phosphatase 1B (PTP1B) inhibitor with an IC50 of 0.25 μM, and without PPARγ agonist activity. KY-226 exerts anti-diabetic and anti-obesity effects by enhancing insulin and leptin signaling, respectively. KY-226 also protects neurons from cerebral ischemic injury. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9601