Learn More
Medchemexpress LLC KY-556 | 110816-78-9 | 725.57 | 25 MG
SDP
Catalog No. 5000448075
Encompass_Preferred
KY-556 is a promising and orally-active pro-agent of disodium cromoglycate (DSCG) that is effective against allergic diseases. It is intended for research use only and appears as a white to off-white solid. This compound has demonstrated anti-allergic effects in various in vivo studies, suggesting its potential as an anti-allergic therapeutic agent.
  • Orally-active pro-agent of disodium cromoglycate (DSCG)
  • Effective against allergic diseases
  • Inhibits 48-hour homologous passive cutaneous anaphylaxis (PCA) in rats
  • Longer duration of action compared to intravenous DSCG
  • Tends to inhibit histamine release from actively sensitized rat lung fragments
  • Prolongs survival time in rat systemic anaphylaxis
  • Significantly inhibits increased airway resistance in experimental asthma in rats
  • Intended for research use only
  • Appears as a white to off-white solid
Catalog No. 50-004-48075 Supplier Medchemexpress LLC Supplier No. 25MGHYU00148
KY-556 is a promising and orally-active pro-agent of disodium cromoglycate (DSCG) that is effective against allergic diseases. It is intended for research use only and appears as a white to off-white solid. This compound has demonstrated anti-allergic effects in various in vivo studies, suggesting its potential as an anti-allergic therapeutic agent.
  • Orally-active pro-agent of disodium cromoglycate (DSCG)
  • Effective against allergic diseases
  • Inhibits 48-hour homologous passive cutaneous anaphylaxis (PCA) in rats
  • Longer duration of action compared to intravenous DSCG
  • Tends to inhibit histamine release from actively sensitized rat lung fragments
  • Prolongs survival time in rat systemic anaphylaxis
  • Significantly inhibits increased airway resistance in experimental asthma in rats
  • Intended for research use only
  • Appears as a white to off-white solid