Learn More
Medchemexpress LLC KY-556 | 110816-78-9 | 725.57 | 50 MG
SDP
Catalog No. 5000449648
Encompass_Preferred
KY-556 is an orally-active pro-agent of disodium cromoglycate (DSCG) with promising efficacy against allergic diseases. It is for research use only.
  • Dose-dependently inhibits 48-hr homologous passive cutaneous anaphylaxis (PCA) in rats.
  • Tends to inhibit histamine release from actively sensitized rat lung fragments.
  • Prolongs survival time in rat systemic anaphylaxis.
  • Significantly inhibits increased airway resistance in experimental asthma in rats.
Catalog No. 50-004-49648 Supplier Medchemexpress LLC Supplier No. HYU0014850MG
KY-556 is an orally-active pro-agent of disodium cromoglycate (DSCG) with promising efficacy against allergic diseases. It is for research use only.
  • Dose-dependently inhibits 48-hr homologous passive cutaneous anaphylaxis (PCA) in rats.
  • Tends to inhibit histamine release from actively sensitized rat lung fragments.
  • Prolongs survival time in rat systemic anaphylaxis.
  • Significantly inhibits increased airway resistance in experimental asthma in rats.