Learn More
Medchemexpress LLC KY-556 (N556) | 110816-78-9 | 725.57 | 10 MG
SDP
Catalog No. 5000448301
Encompass_Preferred
KY-556 is a promising and orally-active pro-agent of disodium cromoglycate (DSCG) against allergic diseases. It is for research use only and not sold to patients.
  • Promising and orally-active pro-agent against allergic diseases
  • Inhibits 48-hr homologous passive cutaneous anaphylaxis (PCA) in rats
  • Longer duration of action compared to intravenous Disodium cromoglycate (DSCG)
  • Inhibits histamine release from actively sensitized rat lung fragments
  • Prolongs survival time in rat systemic anaphylaxis
  • Significantly inhibits increased airway resistance in experimental asthma in rats
Catalog No. 50-004-48301 Supplier Medchemexpress LLC Supplier No. HYU0014810MG
KY-556 is a promising and orally-active pro-agent of disodium cromoglycate (DSCG) against allergic diseases. It is for research use only and not sold to patients.
  • Promising and orally-active pro-agent against allergic diseases
  • Inhibits 48-hr homologous passive cutaneous anaphylaxis (PCA) in rats
  • Longer duration of action compared to intravenous Disodium cromoglycate (DSCG)
  • Inhibits histamine release from actively sensitized rat lung fragments
  • Prolongs survival time in rat systemic anaphylaxis
  • Significantly inhibits increased airway resistance in experimental asthma in rats