Learn More
Medchemexpress LLC HY-131447 5mg Medchemexpress, KY19382 CAS:2226664-93-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1314475MG
Encompass_Preferred
Medchemexpress, HY-131447 5mg KY19382 CAS:2226664-93-1 KY19382 is a potent and orally active dual inhibitor of CXXC5-DVL and GSK3β, with IC50s of 19 and 10 nM, respectively. KY19382 activates Wnt/β-catenin signaling through inhibitory effects on both CXXC5-DVL interaction and GSK3β activity. KY19382 can be used for the research of high fat diet (HFD) induced metabolic diseases. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1283