Learn More
Medchemexpress LLC HY-101090 5mg Medchemexpress, KYA1797K CAS:1956356-56-1 Purity:>98%
SDP
Catalog No. 501932033
Encompass_Preferred
Medchemexpress, HY-101090 5mg KYA1797K CAS:1956356-56-1 KYA1797K is a potent and selective Wnt/β-catenin inhibitor with an IC 50 of 0.75 µM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2033 Supplier Medchemexpress LLC Supplier No. HY1010905MG
Medchemexpress, HY-101090 5mg KYA1797K CAS:1956356-56-1 KYA1797K is a potent and selective Wnt/β-catenin inhibitor with an IC 50 of 0.75 µM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.