Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC KYAT1 Human His 2ug
SDP

Catalog No. 5000196418
Encompass_Preferred

Recombinant human KYAT1 is a His-tagged enzyme expressed in Escherichia coli and supplied lyophilized for stability. It catalyzes the conversion of L-kynurenine to kynurenic acid and is intended for biochemical assays, antibody validation, and protein characterization.

  • Expressed in Escherichia coli with a C-terminal 6xHis tag.
  • Available in small research quantities including 2 μg.
  • Lyophilized formulation in Tris-HCl buffer with trehalose for enhanced stability.
  • Purity greater than 95% as determined by reducing SDS-PAGE.
  • Accession Q16773-2 corresponds to the human sequence (M1-L372).
  • Store frozen; stable at -20 °C for long-term storage and 4 °C after reconstitution for short-term use.

Catalog No. 50-001-96418 Supplier Medchemexpress LLC Supplier No. HYP715332UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human KYAT1 is a His-tagged enzyme expressed in Escherichia coli and supplied lyophilized for stability. It catalyzes the conversion of L-kynurenine to kynurenic acid and is intended for biochemical assays, antibody validation, and protein characterization.

  • Expressed in Escherichia coli with a C-terminal 6xHis tag.
  • Available in small research quantities including 2 μg.
  • Lyophilized formulation in Tris-HCl buffer with trehalose for enhanced stability.
  • Purity greater than 95% as determined by reducing SDS-PAGE.
  • Accession Q16773-2 corresponds to the human sequence (M1-L372).
  • Store frozen; stable at -20 °C for long-term storage and 4 °C after reconstitution for short-term use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.