Learn More
TARGETMOL CHEMICALS INC KYN-101 5MG
SDP
Catalog No. 502877217
Encompass_Preferred
Also available in 1 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. KYN-101 is an orally active and selective aryl hydrocarbon receptor (AHR) inhibitor with anticancer activity. KYN-101 decreases CYP1A1 mRNA expression and can be used in the study of breast cancer and acute myeloid leukemia. KYN-101 is an orally active and selective aryl hydrocarbon receptor (AHR) inhibitor with anticancer activity. Purity 99.39%
Catalog No. 50-287-7217 Supplier TARGETMOL CHEMICALS INC Supplier No. T720565MG
Also available in 1 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. KYN-101 is an orally active and selective aryl hydrocarbon receptor (AHR) inhibitor with anticancer activity. KYN-101 decreases CYP1A1 mRNA expression and can be used in the study of breast cancer and acute myeloid leukemia. KYN-101 is an orally active and selective aryl hydrocarbon receptor (AHR) inhibitor with anticancer activity. Purity 99.39%