Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex Transfer Tubing, Kynar Flex™ 2750 PVDF, 3/16"" ID x 1/4"" OD; 50 ft
Catalog No. NC1676924
Encompass
Applications: High purity and temperature, UV sensitive, or aggressive chemical fluid transfer. Characteristics: Excellent chemical resistance, low TOC, rigid tubing works well with compression and over-molded fittings and adapters. Translucent. Certification: FDA complaint (21 CFR 177.2600), meets USP Class VI requirements, and is made in accordance to cGMP. Temperature range: -40 to 275¿F (-40 to 135¿C) Sterilize: Autoclave, gamma irradiation, or ethylene oxide.
Catalog No. NC1676924 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX9510005
Applications: High purity and temperature, UV sensitive, or aggressive chemical fluid transfer. Characteristics: Excellent chemical resistance, low TOC, rigid tubing works well with compression and over-molded fittings and adapters. Translucent. Certification: FDA complaint (21 CFR 177.2600), meets USP Class VI requirements, and is made in accordance to cGMP. Temperature range: -40 to 275¿F (-40 to 135¿C) Sterilize: Autoclave, gamma irradiation, or ethylene oxide.