Learn More
Cole-Parmer KYNAR GAS SAMPLING BAG, 15X15
Supplier:  Cole-Parmer 0141013
Encompass_Preferred
Cole-Parmer Kynar Gas Sampling Bag, 15" x 15", On/Off valve. Material PVDF. Thickness (mil)3
Catalog No. NC2087305