Learn More
Hach Company PEEK salt bridge with PVDF (Kynar) outer junction - replacement for pHD/pHDsc differential pH/ORP sensors (Hach SB-P1SV)
Catalog No. NC2310683 Shop All Hach Company Products
Encompass
The Hach SB-P1SV PEEK salt bridge with a PVDF (Kynar) outer junction is a field-replaceable reference component for pHD and pHDsc differential pH/ORP sensors. It isolates the sensor's internal reference from process fluids to maintain measurement stability and extend sensor life. Hach recommends replacing the salt bridge every six months for optimum performance.
  • PEEK body with PVDF (Kynar) outer junction for chemical resistance and durability.
  • Field-replaceable double-junction salt bridge for easy maintenance.
  • Compatible with analog and digital pHD differential pH and ORP sensors.
  • Helps maintain stable reference potential and extend sensor service life.
  • Recommended replacement interval: replace every six months for best performance.
Catalog No. NC2310683 Supplier Hach Company Supplier No. SBP1SV
The Hach SB-P1SV PEEK salt bridge with a PVDF (Kynar) outer junction is a field-replaceable reference component for pHD and pHDsc differential pH/ORP sensors. It isolates the sensor's internal reference from process fluids to maintain measurement stability and extend sensor life. Hach recommends replacing the salt bridge every six months for optimum performance.
  • PEEK body with PVDF (Kynar) outer junction for chemical resistance and durability.
  • Field-replaceable double-junction salt bridge for easy maintenance.
  • Compatible with analog and digital pHD differential pH and ORP sensors.
  • Helps maintain stable reference potential and extend sensor service life.
  • Recommended replacement interval: replace every six months for best performance.