Learn More
Sino Biological KYNU Antibody, Rabbit MAb 100uL
Supplier:  Sino Biological 10841R062100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human KYNU (rh KYNU; Catalog#10841-H08B; Q16719; Met1-Asn465).
Catalog No. 50-224-7825