Learn More
Medchemexpress LLC HY-100806 100mg Medchemexpress, Kynurenic acid CAS:492-27-3 Purity:>98%
SDP
Catalog No. 501931955
Encompass_Preferred
Medchemexpress, HY-100806 100mg Kynurenic acid CAS:492-27-3 Kynurenic acid, an endogenous tryptophan metabolite, is a broad-spectrum antagonist targeting NMDA, glutamate, α7 nicotinic acetylcholine receptor . Kynurenic acid is also an agonist of GPR35/CXCR8 . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-1955 Supplier Medchemexpress LLC Supplier No. HY100806100MG
Medchemexpress, HY-100806 100mg Kynurenic acid CAS:492-27-3 Kynurenic acid, an endogenous tryptophan metabolite, is a broad-spectrum antagonist targeting NMDA, glutamate, α7 nicotinic acetylcholine receptor . Kynurenic acid is also an agonist of GPR35/CXCR8 . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.