Learn More
Selleck Chemical LLC Kynurenic acid-100mg
SDP
Catalog No. 502389779
Encompass_Preferred
Kynurenic acid (Quinurenic acid, Kynurenate), a natural metabolite of tryptophan via the kynurenine pathway, is a broad-spectrum excitatory amino acid antagonist; It proved to be an antagonist at NMDA, kainate and AMPAreceptors.
Catalog No. 50-238-9779 Supplier Selleck Chemical LLC Supplier No. S4719100MG
Kynurenic acid (Quinurenic acid, Kynurenate), a natural metabolite of tryptophan via the kynurenine pathway, is a broad-spectrum excitatory amino acid antagonist; It proved to be an antagonist at NMDA, kainate and AMPAreceptors.