Learn More
Abcam Kynurenic acid, endogenous ionotropic / nicotinic antagonist, 1G
Catalog No. 502707893
Encompass_Preferred
MW 189.17 Da, Purity >99%. Endogenous antagonist at ionotropic, glycine β and α7 nicotinic receptors. Neuroprotective *in vivo*. Water-soluble form available - please see Kynurenic acid sodium salt (ab120256).
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7893 Supplier Abcam Supplier No. AB1200641G
MW 189.17 Da, Purity >99%. Endogenous antagonist at ionotropic, glycine β and α7 nicotinic receptors. Neuroprotective *in vivo*. Water-soluble form available - please see Kynurenic acid sodium salt (ab120256).
The product is subject to the following: Abcam Restricted Use Statement