Learn More
Biomatik Corporation Kynurenic Acid (KYNA) Mini sample ELISA Kit
Supplier:  Biomatik Corporation EKU11211
Encompass_Preferred
Kynurenic Acid (KYNA) Mini sample ELISA Kit for detecting KYNA in serum, plasma, other biological fluids. Species Reactivity: Universal. Detection Range: 24.69-2,000ng/ml. Sensitivity: 8.83ng/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 3h. Target Synonyms: Kinurenic acid; Kynuronic acid; Kuinurenic acid; Transtorine. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8197