Learn More
Sigma Aldrich Fine Chemicals Biosciences Human Carcinoma Cells KYSE-410 | CAS N/A | MDL N/A | 1 Vial
Catalog No. NC2315765
Encompass
KYSE-410 cells derived from esophageal squamous cell carcinoma | Origin: 58-year-old male | Cell type: Human carcinoma | Growth: Adherent | Applications: Cancer research, drug screening | Format: 1 vial
Catalog No. NC2315765 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 940720231VL
KYSE-410 cells derived from esophageal squamous cell carcinoma | Origin: 58-year-old male | Cell type: Human carcinoma | Growth: Adherent | Applications: Cancer research, drug screening | Format: 1 vial