Learn More
Sigma Aldrich Fine Chemicals Biosciences KYSE-70 Human Squamous Cell Carcinoma Cell Line | Esophageal Carcinoma Origin | Cisplatin and 5-Fluorouracil Sensitive | In Vitro Esophageal Cancer Model
Catalog No. NC2327929
Encompass
KYSE-70 Human Squamous Cell Carcinoma Cell Line | Esophageal Carcinoma Origin | Cisplatin and 5-Fluorouracil Sensitive | In Vitro Esophageal Cancer Model
Catalog No. NC2327929 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 940720121VL
KYSE-70 Human Squamous Cell Carcinoma Cell Line | Esophageal Carcinoma Origin | Cisplatin and 5-Fluorouracil Sensitive | In Vitro Esophageal Cancer Model