Learn More
Medchemexpress LLC L-006235 | 294623-49-7 | MFCD18086856 | 99.9% | 466.60 | C24H30N6O2S | 1 ML
SDP
Catalog No. 5000245191
Encompass_Preferred
L-006235 (L-235) is a potent, selective, reversible cathepsin K inhibitor sold as an analytical standard for research use. It is supplied as a concentrated DMSO solution or as a solid and is intended for enzymatic assays and analytical characterization.
  • Potent, selective, reversible cathepsin K inhibitor
  • Provided as an analytical standard for research use only
  • Available as a 10 mM solution in DMSO (1 mL) or as a solid (5 mg)
  • High purity (99.93%) and known molecular weight (466.60 g/mol)
  • Suitable for enzymatic assays, bone resorption studies, and analytical reference
Catalog No. 50-002-45191 Supplier Medchemexpress LLC Supplier No. HY10335210MM 1ML
L-006235 (L-235) is a potent, selective, reversible cathepsin K inhibitor sold as an analytical standard for research use. It is supplied as a concentrated DMSO solution or as a solid and is intended for enzymatic assays and analytical characterization.
  • Potent, selective, reversible cathepsin K inhibitor
  • Provided as an analytical standard for research use only
  • Available as a 10 mM solution in DMSO (1 mL) or as a solid (5 mg)
  • High purity (99.93%) and known molecular weight (466.60 g/mol)
  • Suitable for enzymatic assays, bone resorption studies, and analytical reference