Learn More
Medchemexpress LLC L 012 sodium salt | 143556-24-5 | 99.9% | 310.67 | 10 MG
SDP
Catalog No. 5000346893
Encompass_Preferred
L 012 sodium salt is a luminal-based chemiluminescent probe that can detect NADPH oxidase (Nox)-derived superoxide and nitrogen species (reactive oxygen species (ROS) and reactive nitrogen species (RNS)).
  • Luminal-based chemiluminescent probe.
  • Detects NADPH oxidase (Nox)-derived superoxide and nitrogen species (ROS and RNS).
Catalog No. 50-003-46893 Supplier Medchemexpress LLC Supplier No. HY10853750MG
L 012 sodium salt is a luminal-based chemiluminescent probe that can detect NADPH oxidase (Nox)-derived superoxide and nitrogen species (reactive oxygen species (ROS) and reactive nitrogen species (RNS)).
  • Luminal-based chemiluminescent probe.
  • Detects NADPH oxidase (Nox)-derived superoxide and nitrogen species (ROS and RNS).