Learn More
Medchemexpress LLC L 012 (sodium salt) | 143556-24-5 | 99.9% | 310.67 | 25 MG
SDP
Catalog No. 5000347199
Encompass_Preferred
L 012 sodium salt is a luminal-based chemiluminescent probe that can detect NADPH oxidase (Nox)-derived superoxide and nitrogen species (reactive oxygen species (ROS) and reactive nitrogen species (RNS)).
  • Detects NADPH oxidase (Nox)-derived superoxide and nitrogen species.
  • Has a purity of 99.90%.
  • Appears as a solid, white to yellow in color.
  • Recommended storage at 4°C, protected from light, and under nitrogen.
  • In solvent, recommended storage at -80°C for 6 months or -20°C for 1 month, protected from light and under nitrogen.
Catalog No. 50-003-47199 Supplier Medchemexpress LLC Supplier No. 25MGHY108537
L 012 sodium salt is a luminal-based chemiluminescent probe that can detect NADPH oxidase (Nox)-derived superoxide and nitrogen species (reactive oxygen species (ROS) and reactive nitrogen species (RNS)).
  • Detects NADPH oxidase (Nox)-derived superoxide and nitrogen species.
  • Has a purity of 99.90%.
  • Appears as a solid, white to yellow in color.
  • Recommended storage at 4°C, protected from light, and under nitrogen.
  • In solvent, recommended storage at -80°C for 6 months or -20°C for 1 month, protected from light and under nitrogen.