Learn More
TARGETMOL CHEMICALS INC L 012 sodium salt 5MG
SDP
Catalog No. 502877491
Encompass_Preferred
Also available in 1 mL, 1 mg, 10 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. L 012 sodium salt, a luminol analogue, is a widely used chemiluminescent (CL) probe for active oxygen and active nitrogen. L 012 sodium salt is widely used to detect the superoxides (O2•−) derived from NADPH oxidase (Nox) to find Nox inhibitors. Purity 98.34%
Catalog No. 50-287-7491 Supplier TARGETMOL CHEMICALS INC Supplier No. T220965MG
Also available in 1 mL, 1 mg, 10 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. L 012 sodium salt, a luminol analogue, is a widely used chemiluminescent (CL) probe for active oxygen and active nitrogen. L 012 sodium salt is widely used to detect the superoxides (O2•−) derived from NADPH oxidase (Nox) to find Nox inhibitors. Purity 98.34%