Learn More
Selleck Chemical LLC L-(-)-Glucose S3131-25mg
SDP
Catalog No. 503112989
Encompass_Preferred
L-Glucose (L-(-)-Glucose) is an enantiomer of D-glucose L-Glucose promotes food intake while cannot be used by cells as an energy source because it cannot be phosphorylated by hexokinase
Catalog No. 50-311-2989 Supplier Selleck Chemical LLC Supplier No. S313125MG
L-Glucose (L-(-)-Glucose) is an enantiomer of D-glucose L-Glucose promotes food intake while cannot be used by cells as an energy source because it cannot be phosphorylated by hexokinase