Learn More
Selleck Chemical LLC L(-)-Sorbose S5338-25mg
SDP
Catalog No. 503110967
Encompass_Preferred
L(-)-Sorbose (L(-)-Sorbopyranose) is a ketose sugar that is commonly used in the commercial synthesis of ascorbic acid
Catalog No. 50-311-0967 Supplier Selleck Chemical LLC Supplier No. S533825MG
L(-)-Sorbose (L(-)-Sorbopyranose) is a ketose sugar that is commonly used in the commercial synthesis of ascorbic acid