Learn More
ATHENA ES L-15 Leibovitz (with L-glutamine, phenol red, sodium pyruvate), powder (makes 10L)
Catalog No. 5000158091
Encompass_Preferred
L-15 Medium (Leibovitz) is formulated for use in carbon dioxide (CO2) free systems requiring sodium bicarbonate supplements. It supports established cell lines as well as primary explants of embryonic and adult human tissue. L-15 medium can also be used for culturing viruses.
Catalog No. 50-001-58091 Supplier ATHENA ES Supplier No. 0470
L-15 Medium (Leibovitz) is formulated for use in carbon dioxide (CO2) free systems requiring sodium bicarbonate supplements. It supports established cell lines as well as primary explants of embryonic and adult human tissue. L-15 medium can also be used for culturing viruses.