Learn More
ATHENA ES L-15 Leibovitz (with L-glutamine, phenol red, sodium pyruvate), ready-to-use (500mL)
Catalog No. 5000158178
Encompass_Preferred
L-15 Medium (Leibovitz) is formulated for use in carbon dioxide (CO2) free systems requiring sodium bicarbonate supplements. It supports established cell lines as well as primary explants of embryonic and adult human tissue. L-15 medium can also be used for culturing viruses.
Catalog No. 50-001-58178 Supplier ATHENA ES Supplier No. 0471
L-15 Medium (Leibovitz) is formulated for use in carbon dioxide (CO2) free systems requiring sodium bicarbonate supplements. It supports established cell lines as well as primary explants of embryonic and adult human tissue. L-15 medium can also be used for culturing viruses.