Learn More
Quality Biological Inc Leibovitz L-15 Medium 500mL
SDP
Catalog No. 502209731
Encompass_Preferred
Most media are buffered by a combination of sodium bicarbonate and an atmosphere enriched in CO2. The latter is usually provided through the use of a specially equipped incubator. L-15 was developed at a time when such equipment was not readily available. Dr. Leibovitz developed a buffering system based on free base amino acids inorganic salts and the substitution of galactose for glucose. Without sodium bicarbonateDesigned for cell culture without CO2With pheonl redWithout L-glutamine Manufactured in an ISO 90012015 certified facility.
Catalog No. 50-220-9731 Supplier Quality Biological Inc Supplier No. 112029101
Most media are buffered by a combination of sodium bicarbonate and an atmosphere enriched in CO2. The latter is usually provided through the use of a specially equipped incubator. L-15 was developed at a time when such equipment was not readily available. Dr. Leibovitz developed a buffering system based on free base amino acids inorganic salts and the substitution of galactose for glucose. Without sodium bicarbonateDesigned for cell culture without CO2With pheonl redWithout L-glutamine Manufactured in an ISO 90012015 certified facility.