Learn More
Worthington Biochemical Corporation L-15 MEDIA POWDER (L15NK)
SDP
Catalog No. 50592502
Encompass
Leibovitz L-15 media powder, a component of the NCIS kit. Reconstitute entire contents of pouch, QS to 1 liter with cell culture grade water, and 0.22 micron filter. Suitable for cell isolation and culture applications.
Catalog No. 50-592-502 Supplier Worthington Biochemical Corporation Supplier No. LK003250
Leibovitz L-15 media powder, a component of the NCIS kit. Reconstitute entire contents of pouch, QS to 1 liter with cell culture grade water, and 0.22 micron filter. Suitable for cell isolation and culture applications.