Learn More
Sigma Aldrich Fine Chemicals Biosciences L-15 Medium (Leibovitz), 500 mL
Catalog No. NC1733735
Encompass
Originally formulated for use in carbon dioxide free systems requiring sodium bicarbonate supplement. L-15 supports established cell lines as well as primary explants of embryonic and adult human tissue.
Catalog No. NC1733735 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. L1518500ML
Originally formulated for use in carbon dioxide free systems requiring sodium bicarbonate supplement. L-15 supports established cell lines as well as primary explants of embryonic and adult human tissue.