Learn More
Medchemexpress LLC HY-107734 10mg Medchemexpress, L 152804 CAS:6508-43-6 Purity:>98%
SDP
Catalog No. 501968458
Encompass_Preferred
Medchemexpress, HY-107734 10mg L 152804 CAS:6508-43-6 L 152804 is an orally active and selective neuropeptide Y Y5 receptor (NPY5-R) antagonist, with a Ki of 26 nM for hY5. L 152804 causes weight loss in diet-induced obese mice by modulating food intake and energy expenditure. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8458 Supplier Medchemexpress LLC Supplier No. HY10773410MG
Medchemexpress, HY-107734 10mg L 152804 CAS:6508-43-6 L 152804 is an orally active and selective neuropeptide Y Y5 receptor (NPY5-R) antagonist, with a Ki of 26 nM for hY5. L 152804 causes weight loss in diet-induced obese mice by modulating food intake and energy expenditure. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.