Learn More
Cayman Chemical L-161,982, 147776-06-5, 1 mg
SDP
Supplier:  Cayman Chemical 100115651
Encompass_Preferred
A potent and selective EP4 receptor antagonist (Ki = 24 nM); reverses the anti-inflammatory action of PGE2 in LPS - activated human macrophages at 100 nM
Catalog No. NC1026095