Learn More
TARGETMOL CHEMICALS INC L-161240 5MG
SDP
Catalog No. 502580330
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. L-161240 is an antimicrobial agent associated with lipid A biosynthesis with an IC50 value of 30 nM as determined by DEACET and a MIC value of 1-3 ug/ mL against wild-type E. coli. purity: 97%
Catalog No. 50-258-0330 Supplier TARGETMOL CHEMICALS INC Supplier No. T324905MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. L-161240 is an antimicrobial agent associated with lipid A biosynthesis with an IC50 value of 30 nM as determined by DEACET and a MIC value of 1-3 ug/ mL against wild-type E. coli. purity: 97%