Learn More
Medchemexpress LLC HY-20019 5mg Medchemexpress, L-165041 CAS:79558-09-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY200195MG
Encompass_Preferred
Medchemexpress, HY-20019 5mg L-165041 CAS:79558-09-1 L-165041 is a cell permeable PPARδ agonist, with Kis of 6 nM and appr 730 nM for PPARδ and PPARγ, respectively, and induces adipocyte differentiation in NIH-PPARδ cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2213