Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-2,4-Diaminobutyric acid dihydrochloride, 98%, Thermo Scientific™

Catalog No. AC218090050
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC218090050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC218090050 Supplier Thermo Scientific Chemicals Supplier No. 218090050

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1883-09-6
Molecular Formula C4H12Cl2N2O2
Molecular Weight (g/mol) 191.052
MDL Number MFCD00064561
InChI Key CKAAWCHIBBNLOJ-QTNFYWBSSA-N
Synonym l-2,4-diaminobutyric acid dihydrochloride, s-2,4-diaminobutanoic acid dihydrochloride, s-+-2,4-diaminobutyric acid dihydrochloride, h-dab-oh.2hcl, 2,4-diaminobutyric acid dihydrochloride, 2s-2,4-diaminobutanoic acid dihydrochloride, s-+-2,4-diamino-n-butyric acid dihydrochloride, butanoic acid, 2,4-diamino-, dihydrochloride, 2s, d-dab.2hcl, 2s-2,4-diaminobutanoic acid, chloride, chloride
PubChem CID 2724265
IUPAC Name (2S)-2,4-diaminobutanoic acid;dihydrochloride
SMILES C(CN)C(C(=O)O)N.Cl.Cl

Specifications

Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Linear Formula H2NCH2CH2CH(NH2)CO2H·2HCl
Quantity 5 g
Beilstein 04, II, 839
Specific Rotation Condition +14° (16°C c=3.7,H2O)
Specific Rotation +14° to +16° (20°C, 589nm) (c=3.7, H2O)
Formula Weight 191.06
Percent Purity ≥97.5%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material L-2, 4-Diaminobutyric acid dihydrochloride, 98%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.