Learn More
Medchemexpress LLC L-2,4-diaminobutyric acid hydrobromide | 73143-97-2 | MFCD07368664 | 98.0% | 199.05 g/mol | C4H11BrN2O2 | 500 MG
SDP
Catalog No. 5000249122
Encompass_Preferred
L-2,4-diaminobutyric acid hydrobromide (L-DABA hydrobromide) is a research chemical described as a weak GABA transaminase inhibitor (IC50 > 500 μM) and reported to exhibit antitumor activity in vitro and in vivo. It is supplied as the hydrobromide salt, CAS 73143-97-2, with molecular weight 199.05 g/mol and reported purity 98.0%.
  • High purity: 98.0%.
  • Reported activity: weak GABA transaminase inhibitor (IC50 > 500 μM).
  • Demonstrated antitumor activity in vitro and in vivo.
  • Provided as hydrobromide salt for research use.
  • Available in 500 mg pack size.
Catalog No. 50-002-49122 Supplier Medchemexpress LLC Supplier No. HY101414A500MG
L-2,4-diaminobutyric acid hydrobromide (L-DABA hydrobromide) is a research chemical described as a weak GABA transaminase inhibitor (IC50 > 500 μM) and reported to exhibit antitumor activity in vitro and in vivo. It is supplied as the hydrobromide salt, CAS 73143-97-2, with molecular weight 199.05 g/mol and reported purity 98.0%.
  • High purity: 98.0%.
  • Reported activity: weak GABA transaminase inhibitor (IC50 > 500 μM).
  • Demonstrated antitumor activity in vitro and in vivo.
  • Provided as hydrobromide salt for research use.
  • Available in 500 mg pack size.