Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-2-Aminoadipic Acid 97.0+%, TCI America™
SDP

Catalog No. A14575G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A14575G 5 g
A14571G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A14575G Supplier TCI America Supplier No. A14575G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1118-90-7
Molecular Formula C6H11NO4
Molecular Weight (g/mol) 161.16
MDL Number MFCD00002636
InChI Key OYIFNHCXNCRBQI-BYPYZUCNSA-N
Synonym l-2-aminoadipic acid, s-2-aminohexanedioic acid, 2s-2-aminohexanedioic acid, l-alpha-aminoadipic acid, l-alpha-aminoadipate, l-2-amino adipic acid, l-homoglutamic acid, aminoadipate, l-2-aminoadipate, homoglutamic acid
PubChem CID 92136
ChEBI CHEBI:37023
IUPAC Name (2S)-2-aminohexanedioic acid
SMILES N[C@@H](CCCC(O)=O)C(O)=O

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 161.16
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material L-2-Aminoadipic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.