Learn More
Medchemexpress LLC L-2-Aminobutyric acid | 1492-24-6 | 103.12 | 1 ML
SDP
Catalog No. 5000366529
Encompass_Preferred
L-2-Aminobutyric acid is a non-proteinogenic amino acid found in human tissues and body fluids, formed through the transamination of oxobutyric acid. It can act as a precursor for anticonvulsant and anti-tuberculosis agents and is a crucial intermediate in the chemical and pharmaceutical industries. Elevated levels in plasma are associated with conditions such as Reye's syndrome, tyrosinemia, homocystinuria, nonketotic hyperglycinemia, and ornithine transcarbamylase deficiency.
  • Non-proteinogenic amino acid
  • Found in human tissues and body fluids
  • Formed via transamination of oxobutyric acid
  • Precursor for anticonvulsant and anti-tuberculosis agents
  • Crucial intermediate in chemical and pharmaceutical industries
  • Elevated levels associated with specific medical conditions
Catalog No. 50-003-66529 Supplier Medchemexpress LLC Supplier No. HYW01058910MM/1ML
L-2-Aminobutyric acid is a non-proteinogenic amino acid found in human tissues and body fluids, formed through the transamination of oxobutyric acid. It can act as a precursor for anticonvulsant and anti-tuberculosis agents and is a crucial intermediate in the chemical and pharmaceutical industries. Elevated levels in plasma are associated with conditions such as Reye's syndrome, tyrosinemia, homocystinuria, nonketotic hyperglycinemia, and ornithine transcarbamylase deficiency.
  • Non-proteinogenic amino acid
  • Found in human tissues and body fluids
  • Formed via transamination of oxobutyric acid
  • Precursor for anticonvulsant and anti-tuberculosis agents
  • Crucial intermediate in chemical and pharmaceutical industries
  • Elevated levels associated with specific medical conditions