Learn More
Medchemexpress LLC HY-W015114 10mg Medchemexpress, L-2-Hydroxyglutaric acid (disodium) CAS:63512-50-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYW01511410MG
Encompass_Preferred
Medchemexpress, HY-W015114 10mg L-2-Hydroxyglutaric acid (disodium) CAS:63512-50-5 L-2-Hydroxyglutaric acid disodium is an epigenetic modifier and putative oncometabolite in renal cancer. L-2-Hydroxyglutaric acid disodium can inhibit histone demethylases and hence promote histone methylation. L-2-Hydroxyglutaric acid inhibits mitochondrial creatine kinase (Mi-CK) activity with Km and Ki of 2.52 mM and 11.13 mM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3369